Potassium-3,5-bis(trifluoromethyl)phenyltrifluoroborate 96%

Supplier: Thermo Scientific
Warning

H53113.06EA 201 EUR
H53113.06
Potassium-3,5-bis(trifluoromethyl)phenyltrifluoroborate 96%
Potassium 3,5-bis(trifluoromethyl)phenyltrifluoroborate
Formula: C₈H₃BF₉K
MW: 320,01
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03701625
CAS Number: 166328-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR