2-Benzo[b]thiophenesulphonyl chloride

MAYBCC12203CBEA 0 EUR
MAYBCC12203CB MAYBCC12203DA
2-Benzo[b]thiophenesulphonyl chloride
2-Benzo[b]thiophenesulphonyl chloride
Formula: C₈H₅ClO₂S₂
MDL Number: MFCD03659701
CAS Number: 90001-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR