2,4-Dibromo-N-(4-fluorobenzyl)aniline, 97%

H57336.MDEA 72.3 EUR
ALFAH57336.MD ALFAH57336.03
2,4-Dibromo-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1152667-75-8
Molecular Formula: C13H10Br2FN
Formula Weight: 359.03
MDL No.: MFCD12566003
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR