4-Bromo-2-fluoro-N-(4-fluorobenzyl)aniline, 97%

H57864.MDEA 402.5 EUR
ALFAH57864.MD ALFAH57864.03
4-Bromo-2-fluoro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019614-02-8
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
MDL No.: MFCD11142749
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR