2-Bromo-4-fluoro-N-(3-fluorobenzyl)aniline, 97%

H57292.03EA 0 EUR
ALFAH57292.03 ALFAH57292.MD
2-Bromo-4-fluoro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1038978-91-4
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
MDL No.: MFCD11189236
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR