2,3-Dichloro-N-(3-fluorobenzyl)aniline, 97%

H57462.03EA 0 EUR
ALFAH57462.03 ALFAH57462.MD
2,3-Dichloro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1036454-45-1
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD11119388
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR