2,5-Dichloro-N-(4-fluorobenzyl)aniline, 97%

H57154.MDEA 0 EUR
ALFAH57154.MD ALFAH57154.03
2,5-Dichloro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 940355-99-7
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD11058526
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR