4-Chloro-2-fluoro-N-(3-fluorobenzyl)aniline, 97%

H57731.MDEA 45.1 EUR
ALFAH57731.MD ALFAH57731.03
4-Chloro-2-fluoro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1152635-39-6
Molecular Formula: C13H10ClF2N
Formula Weight: 253.67
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR