N-(2-Fluorobenzyl)-2-iodoaniline, 97%

H57097.MDEA 0 EUR
ALFAH57097.MD ALFAH57097.03
N-(2-Fluorobenzyl)-2-iodoaniline, 97%
Purity: 97%
CAS No.: 376605-69-5
Molecular Formula: C13H11FIN
Formula Weight: 327.14
MDL No.: MFCD02162651
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR