2,6-Difluoro-N-(3-fluorobenzyl)aniline, 97%

H57261.03EA 289.7 EUR
ALFAH57261.03 ALFAH57261.MD
2,6-Difluoro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1039839-41-2
Molecular Formula: C13H10F3N
Formula Weight: 237.22
MDL No.: MFCD11180926
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR