Lead antimonide ≥99% (metals basis)
|
Danger
|
36350.14EA
409
EUR
36350.14
36350.06
Lead antimonide ≥99% (metals basis)
Lead antimonide
Formula:
PbSb MW: 328,96 g/mol Density: 6,72 Storage Temperature: Ambient |
MDL Number:
MFCD00151190 CAS Number: 12266-38-5 EINECS: 235-536-6 UN: 1549 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...