Gadolinium disilicide ≥99.9% (metals basis)
Synonyms:
Gadolinium silicide
36356.09EA
544
EUR
36356.09
36356.04
Gadolinium disilicide ≥99.9% (metals basis)
Gadolinium disilicide
Formula:
GdSi₂ MW: 213,42 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00144382 CAS Number: 12134-75-7 EINECS: 235-216-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...