Pentacopper silicide ≥99.5% (metals basis)
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Copper silicide
12844.22EA
365
EUR
12844.22
12844.14
Pentacopper silicide ≥99.5% (metals basis)
Pentacopper silicide
Formula:
Cu₅Si MW: 345,79 g/mol Melting Pt: 825 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134719 CAS Number: 12159-07-8 EINECS: 235-286-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...