Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
|
Danger
|
Synonyms:
Chromium silicide
41060.KSEA
635
EUR
41060.KS
Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Chromium disilicide
Magnetron sputtering source
Formula:
CrSi₂ MW: 108,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00168070 CAS Number: 12018-09-6 EINECS: 234-633-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...