Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
|
Danger
|
Synonyms:
Chromium silicide
41061.KSEA
691
EUR
41061.KS
Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Chromium disilicide
Formula:
CrSi₂ MW: 108,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00168070 CAS Number: 12018-09-6 EINECS: 234-633-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...