Manganese antimonide ≥99.5% (metals basis)
|
Warning
|
36385.06EA
121
EUR
36385.06
Manganese antimonide ≥99.5% (metals basis)
Manganese antimonide
Formula:
MnSb |
MDL Number:
MFCD00016215 CAS Number: 12032-82-5 EINECS: 234-779-5 UN: 1549 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...