Triiron aluminide ≥97.5% (metals basis)
Synonyms:
Iron aluminide
40006.18EA
104
EUR
40006.18
Triiron aluminide ≥97.5% (metals basis)
Triiron aluminide
Applications: Substitute for stainless steel with better high temperature strength. Good corrosion resistance in sulfidizing environments
Formula:
AlFe₃ Melting Pt: 1540 °C Density: 6,72 |
MDL Number:
MFCD00210617 CAS Number: 12003-42-8 EINECS: 234-427-0 UN: 3178 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...