4,7-Dibromo-3-hydroxy-2-naphthoic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid
L07019.14EA
217
EUR
L07019.14
4,7-Dibromo-3-hydroxy-2-naphthoic acid ≥97%
4,7-Dibromo-3-hydroxy-2-naphthoic acid
Formula:
C₁₁H₆Br₂O₃ MW: 345,98 g/mol Melting Pt: 249…253 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046367 CAS Number: 1779-10-8 EINECS: 217-214-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...