4-Bromo-1-fluoronaphthalene-2-boronic acid, 98%

H52911.03EA 151.3 EUR
ALFAH52911.03 ALFAH52911.MD
4-Bromo-1-fluoronaphthalene-2-boronic acid, 98%
Purity: 98%
CAS No.: 913836-09-6
Molecular Formula: C10H7BBrFO2
Formula Weight: 268.88
Melting Point: >300°
MDL No.: MFCD09027236
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR