2-Chloro-6-methylisonicotinic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Chloro-6-methylpyridine-4-carboxylic acid
L10544.06EA
213
EUR
L10544.06
L10544.03
2-Chloro-6-methylisonicotinic acid ≥98%
2-Chloro-6-methylisonicotinic acid
Formula:
C₇H₆ClNO₂ MW: 171,58 g/mol Melting Pt: ∼ 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052830 CAS Number: 25462-85-5 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...