3-Aminothiophene-2-carboxamide ≥97%

Supplier: Thermo Scientific

L12371.MDEA 39 EUR
L12371.MD L12371.03
3-Aminothiophene-2-carboxamide ≥97%
3-Aminothiophene-2-carboxamide
Formula: C₅H₆N₂OS
MW: 142,18
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00052593
CAS Number: 147123-47-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR