2,5-Dimethyl-3-thiophenesulphonyl chloride

MAYBMO00053DAEA 0 EUR
MAYBMO00053DA
2,5-Dimethyl-3-thiophenesulphonyl chloride
2,5-Dimethyl-3-thiophenesulphonyl chloride
Formula: C₆H₇ClO₂S₂
MDL Number: MFCD03086201
CAS Number: 97272-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR