2,1,3-Benzothiadiazole-5-carbaldehyde

MAYBCC09204CBEA 0 EUR
MAYBCC09204CB MAYBCC09204DA
2,1,3-Benzothiadiazole-5-carbaldehyde
2,1,3-Benzothiadiazole-5-carbaldehyde
Formula: C₇H₄N₂OS
MDL Number: MFCD00457949
CAS Number: 71605-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR