4-Amino-2-chloro-6,7-dimethoxyquinazoline
Supplier: Molekula
Synonyms:
2-Chloro-4-amino-6,7-dimethoxyquinazoline
MOLEM31299379EA
0
EUR
MOLEM31299379
4-Amino-2-chloro-6,7-dimethoxyquinazoline
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Formula:
C₁₀H₁₀ClN₃O₂ MW: 239,66 g/mol |
MDL Number:
MFCD00051734 CAS Number: 23680-84-4 EINECS: 245-821-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...