2,4-Diaminoquinazoline ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Quinazoline-2,4-diamine
L13783.03EA
71.4
EUR
L13783.03
L13783.06
2,4-Diaminoquinazoline ≥98%
2,4-Diaminoquinazoline
Formula:
C₈H₈N₄ MW: 160,18 g/mol Melting Pt: 254…258 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236032 CAS Number: 1899-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...