1-Amino-2-methyl-9,10-anthraquinone, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

42994-25MGEA 37.4 EUR
42994-25MG
1-Amino-2-methyl-9,10-anthraquinone, Supelco®
1-Amino-2-methyl-9,10-anthraquinone
Formula: C₁₅H₁₁NO₂
MDL Number: MFCD00001220
CAS Number: 82-28-0
EINECS: 201-408-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR