Potassium-4-(1-piperidinylcarbonyl)phenyltrifluoroborate ≥97%

Supplier: Thermo Scientific

Warning

Synonyms: Potassium trifluoro[4-(piperidine-1-carbonyl)phenyl]boranuide

H51146.06EA 147 EUR
H51146.06 H51146.03
Potassium-4-(1-piperidinylcarbonyl)phenyltrifluoroborate ≥97%
Potassium 4-(1-piperidinylcarbonyl)phenyltrifluoroborate
Formula: C₁₂H₁₄BF₃KNO
MW: 295,15
Storage Temperature: Ambient
MDL Number: MFCD11977715
CAS Number: 1359865-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR