2-(Difluoromethoxy)-5-fluorobenzonitrile 95%, JRD

Supplier: Thermo Scientific

Danger

H31695.MDEA 53.1 EUR
H31695.MD H31695.03
2-(Difluoromethoxy)-5-fluorobenzonitrile 95%, JRD
2-(Difluoromethoxy)-5-fluorobenzonitrile
Formula: C₈H₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832329
CAS Number: 1017778-48-1
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR