3-Fluoro-5-(trifluoromethoxy)benzonitrile ≥97%

Supplier: Thermo Scientific
Danger

H33944.06EA 283 EUR
H33944.06 H33944.03
3-Fluoro-5-(trifluoromethoxy)benzonitrile ≥97%
3-Fluoro-5-(trifluoromethoxy)benzonitrile
Formula: C₈H₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD18783224
CAS Number: 1352999-93-9
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR