4-(Difluoromethoxy)-2-fluorobenzonitrile 97%, JRD

Supplier: Thermo Scientific

Danger

H31690.03EA 141 EUR
H31690.03 H31690.MD
4-(Difluoromethoxy)-2-fluorobenzonitrile 97%, JRD
4-(Difluoromethoxy)-2-fluorobenzonitrile
Formula: C₈H₄F₃NO
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD09832366
CAS Number: 1017779-49-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR