3-[5-Fluoro-2-(trifluoromethoxy)phenyl]propionic acid, JRD, 97%

H32019.03EA 94.1 EUR
ALFAH32019.03 ALFAH32019.MD
3-[5-Fluoro-2-(trifluoromethoxy)phenyl]propionic acid, JRD, 97%
Purity: 97%
Molecular Formula: C10H8F4O3
Formula Weight: 252.16
Refractive Index: 1.4490
MDL No.: MFCD11519341
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR