3-(4-Ethoxy-3-(trifluoromethyl)phenyl)propionic acid ≥97%

Supplier: Thermo Scientific

Warning

H33980.MDEA 60 EUR
H33980.MD H33980.03
3-(4-Ethoxy-3-(trifluoromethyl)phenyl)propionic acid ≥97%
3-(4-Ethoxy-3-(trifluoromethyl)phenyl)propionic acid
Formula: C₁₂H₁₃F₃O₃
MW: 262,23
Storage Temperature: Ambient
MDL Number: MFCD13196589
CAS Number: 1206593-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR