3-(4-Fluoro-3-(trifluoromethoxy)phenyl)propionic acid ≥97%

Supplier: Alfa Aesar
Warning

H34036.03EA 110 EUR
H34036.03 H34036.MD
3-(4-Fluoro-3-(trifluoromethoxy)phenyl)propionic acid ≥97%
3-(4-Fluoro-3-(trifluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₈F₄O₃
MW: 252,16
Storage Temperature: Ambient
MDL Number: MFCD09999449
CAS Number: 1240256-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR