2,3-Difluoro-p-toluidine ≥97%

Supplier: Thermo Scientific

Danger

Synonyms: 2,3-Difluoro-4-methylaniline

H26731.06EA 301 EUR
H26731.06 H26731.03
2,3-Difluoro-p-toluidine ≥97%
2,3-Difluoro-p-toluidine
Formula: C₇H₇F₂N
MW: 143,14
Storage Temperature: Ambient
MDL Number: MFCD06660321
CAS Number: 886503-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR