5-Amino-2,4-difluorophenol 98%
|
Danger
|
H32479.06EA
277
EUR
H32479.06
5-Amino-2,4-difluorophenol 98%
5-Amino-2,4-difluorophenol
Formula:
C₆H₅F₂NO MW: 145,11 g/mol Melting Pt: ∼ 177 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04973718 CAS Number: 113512-71-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...