2-Methyl-3-(trifluoromethyl)anisole ≥97%

Supplier: Alfa Aesar
Warning

H32706.03EA 175 EUR
H32706.03 H32706.MD
2-Methyl-3-(trifluoromethyl)anisole ≥97%
2-Methyl-3-(trifluoromethyl)anisole
Formula: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD09832284
CAS Number: 1017778-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR