Aluminium antimonide ≥99.99% (metals basis)

Supplier: Thermo Scientific

Warning

43455.14EA 765 EUR
43455.14 43455.06
Aluminium antimonide ≥99.99% (metals basis)
Aluminium antimonide

In semiconductor technology

Formula: AlSb
MW: 148,73
Melting Pt: 1050 °C
Storage Temperature: Ambient
MDL Number: MFCD00014154
CAS Number: 25152-52-7
EINECS: 246-667-3
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR