D-(-)-α-(4-Hydroxyphenyl)glycine

Supplier: Molekula

Synonyms: H-D-Phg(4-OH)-OH(-)-4-Hydroxy-D-phenylglycine(R)-(-)-2-(4-Hydroxyphenyl)glycineD-(-)-p-HydroxyphenylglycineD(-)-4-Hydroxyphenylglycine

MOLEM57712989EA 0 EUR
MOLEM57712989
D-(-)-α-(4-Hydroxyphenyl)glycine
D(-)-α-(4-Hydroxyphenyl)glycine
Formula: C₈H₉NO₃
MW: 167,16 g/mol
MDL Number: MFCD00004262
CAS Number: 22818-40-2
EINECS: 245-247-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR