3,6-Difluorosalicylic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,6-Difluoro-2-hydroxybenzoic acid

H32209.MDEA 73.8 EUR
H32209.MD H32209.03
3,6-Difluorosalicylic acid ≥97%
3,6-Difluorosalicylic acid
Formula: C₇H₄F₂O₃
MW: 174,10
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD08275285
CAS Number: 749230-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR