4-Bromo-3-(trifluoromethyl)benzoic acid ≥98%

Warning

H31618.03EA 57 EUR
H31618.03 H31618.06
4-Bromo-3-(trifluoromethyl)benzoic acid ≥98%
4-Bromo-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08459288
CAS Number: 161622-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR