3-Methoxy-4-(trifluoromethyl)benzoic acid ≥97%
|
Warning
|
H33574.06EA
334
EUR
H33574.06
H33574.03
3-Methoxy-4-(trifluoromethyl)benzoic acid ≥97%
3-Methoxy-4-(trifluoromethyl)benzoic acid
Formula:
C₉H₇F₃O₃ MW: 220,15 g/mol Melting Pt: 209…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD13185682 CAS Number: 276861-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...