2-Methyl-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: Thermo Scientific
Warning

H26564.09EA 208 EUR
H26564.09 H26564.03
2-Methyl-5-(trifluoromethyl)benzoic acid ≥97%
2-Methyl-5-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD01631589
CAS Number: 13055-63-5
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR