Triethylphosphine oxide

Supplier: Thermo Scientific
Warning

30391.03EA 67.8 EUR
30391.03 30391.06
Triethylphosphine oxide
Triethylphosphine oxide
Formula: C₆H₁₅OP
MW: 134,16 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00013914
CAS Number: 597-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR