2,3-Difluoroisonicotinic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,3-Difluoropyridine-4-carboxylic acid
H33463.03EA
81.9
EUR
H33463.03
2,3-Difluoroisonicotinic acid ≥97%
2,3-Difluoroisonicotinic acid
Formula:
C₆H₃F₂NO₂ MW: 159,09 g/mol Melting Pt: 165…167 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09031174 CAS Number: 851386-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...