3-Chloro-2-fluoro-6-(trifluoromethyl)benzonitrile 97%, JRD
|
Danger
|
XXXXJRD-2545.5G
3-Chloro-2-fluoro-6-(trifluoromethyl)benzonitrile 97%, JRD
3-Chloro-2-fluoro-6-(trifluoromethyl)benzonitrile
Formula:
C₈H₂ClF₄N MW: 223,55 g/mol Melting Pt: 46…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD11519382 CAS Number: 186517-00-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...