2-Chloro-4-(trifluoromethyl)anisole ≥97%

Supplier: Thermo Scientific

Warning

H33554.MDEA 53.1 EUR
H33554.MD H33554.03
2-Chloro-4-(trifluoromethyl)anisole ≥97%
2-Chloro-4-(trifluoromethyl)anisole
Formula: C₈H₆ClF₃O
MW: 210,58
Storage Temperature: Ambient
MDL Number: MFCD13185462
CAS Number: 1214340-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR