3-Methyl-5-(trifluoromethoxy)benzonitrile ≥97%

Supplier: Thermo Scientific
Danger

H31921.03EA 53.3 EUR
H31921.03 H31921.06
3-Methyl-5-(trifluoromethoxy)benzonitrile ≥97%
3-Methyl-5-(trifluoromethoxy)benzonitrile
Formula: C₉H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09025378
CAS Number: 916420-59-2
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR