4-Ethoxy-3-(trifluoromethyl)benzonitrile ≥97%

Danger

H33218.06EA 140 EUR
H33218.06 H33218.03
4-Ethoxy-3-(trifluoromethyl)benzonitrile ≥97%
4-Ethoxy-3-(trifluoromethyl)benzonitrile
Formula: C₁₀H₈F₃NO
MW: 215,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13196579
CAS Number: 1206593-22-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR