2,3-Dichloro-6-fluorobenzonitrile ≥97%

Supplier: Thermo Scientific
Danger

H26584.03EA 64.8 EUR
H26584.03 H26584.06
2,3-Dichloro-6-fluorobenzonitrile ≥97%
2,3-Dichloro-6-fluorobenzonitrile
Formula: C₇H₂Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD04115878
CAS Number: 79544-26-6
EINECS: None
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR