3,6-Dichloro-2-fluorobenzonitrile ≥97%
|
Danger
|
H32752.06EA
216
EUR
H32752.06
H32752.03
3,6-Dichloro-2-fluorobenzonitrile ≥97%
3,6-Dichloro-2-fluorobenzonitrile
Formula:
C₇H₂Cl₂FN MW: 190,00 g/mol Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD09025385 CAS Number: 916420-65-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...